Order Entry
Italy
ContactUsLinkComponent
2,3-Dimethoxyphenol 99%
Irritant
2,3-Dimethoxyphenol 99%
Certificates
undefined
Irritant
2,3-Dimethoxyphenol 99%

Specification Test Results

  • Appearance:
    Clear orange to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5385 to 1.5405 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together