Wright's stain used as a blood stain, Sigma-Aldrich®
:68988-92-1
:51938-72-8
:MFCD00082143
:0.8 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:273-541-5
:GHS07
:Room Temperature
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Solubility (Colour)
c = 100 mg/100 ml; Methanol:Purple to Very Dark Purple to Very Dark Blue and Black - Solubility (Turbidity):Clear to Turbid
- Assay (HPLC)
Impurity: Azure C:≤4 % - Suitability
Suitable for use in blood staining:Suitable
Frequently Bought Together