1,4-Dimethoxybenzene 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:150-78-7
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00008401
:138.17 g/mol
Boiling Pt:212.6 °C (1013 hPa)
Melting Pt:54…58 °C
Density:1.05 g/cm³
Flash Pt:98 °C
EINECS:205-771-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white to grey Crystals of flakes
- Infrared spectrum:Conforms
- Melting point:54 to 58 °C
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together