
Dimethyl-1,5-naphthalenedisulphonate ≥98.0% (by GC)
TCIAN0439-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:200.0 to 204.0 deg-C
- Solubility in hot Toluene:almost transparency
:
:316.34 g/mol
:110 °C
:201-317-9
:8,III
:GHS05
:20779-13-9
:C₁₂H₁₂O₆S₂
:MFCD00059544