


3-Pyrroline tech. 85% (remainder pyrrolidine), Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-96-6
:
Formula:C₄H₇N
MDL Number:MFCD00005213
:69.11 g/mol
Merck Index:14,8017
Boiling Pt:90…91 °C
Melting Pt:–71 °C
Density:0.902
Flash Pt:–18 °C (0 °F)
Storage Temperature:Refrigerator
EINECS:203-723-1
UN:2733
ADR:3,II
:GHS02
:GHS05
:GHS07
Related Information
Certificates
Search by Lot Numbermsds_ALFAH26072
Scheda di sicurezza
Organic Building Blocks and Intermediates
Note applicative
Frequently Bought Together