Order Entry
Italy
ContactUsLinkComponent
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3008600
Environment
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3008600
Catalog # REGE3008600
Supplier:  REAGECON
CAS Number:  
Environment
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3008600
Catalog # REGE3008600
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3082
ADR:9,III
 :GHS09
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302