Dimethyl sebacate
Catalog # EHERC15711030
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:230.3 g/mol
Boiling Pt:102 °C (0.05 mmHg)
Melting Pt:38 °C
Density:1.0989 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-431-4
CAS Number:106-79-6
Formula:C₁₂H₂₂O₄
MDL Number:MFCD00008472