
Methanesulphonic anhydride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7143-01-3
:
Formula:C₂H₆O₅S₂
MDL Number:MFCD00007556
:174.2 g/mol
Boiling Pt:125 °C (4 mmHg)
Melting Pt:69…70 °C
Storage Temperature:Ambient
EINECS:230-442-1
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to grey to light brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Titration by coupling:≥96.0 %
Specification Test Results
Frequently Bought Together