
Anisole for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-66-3
:
Formula:C₇H₈O
MDL Number:MFCD00008354
:108.14 g/mol
Merck Index:13,00672
Boiling Pt:154 °C (1013 hPa)
Melting Pt:–37 °C
Density:0.9961 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-876-1
UN:2222
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:44 °C
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Solubility:1.6 g/l (25 °C)
- Melting Point:-37 °C
- Molar Mass:108.13 g/mol
- Boiling Point:156 °C (1013 hPa)
- Vapor Pressure:3.2 hPa (20 °C)
- Flash Point:44 °C
- Explosion Limit:0.34 - 6.3 %(V)
- Density:0.994 g/cm3 (20 °C)
- pH:(H2O, 20 °C) not applicable
- Ignition Point:475 °C
Specification Test Results
Related Information
Frequently Bought Together