4-Nitrophenol ≥99% (by GC), crystalline powder for spectroscopy
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:100-02-7
:
Formula:C₆H₅NO₃
MDL Number:MFCD00007331
:139.11 g/mol
Merck Index:13,06654
Boiling Pt:279 °C (1013 hPa)
Melting Pt:105…115 °C
Density:1.48 g/cm³ (20 °C)
Flash Pt:169 °C
Storage Temperature:Refrigerator
EINECS:202-811-7
UN:1663
ADR:6.1,III
:GHS08
:GHS07
Specification Test Results
- Identity Test :Passes
- Purity:≥99% (GC)
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together