Media bottles, graduated, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
Amber borosilicate glass bottle with graduations and marking patch.
- Graduations and marking patch assists in measuring volume and identifying content
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
Ordering information: Black phenolic screw cap can be purchased separately
Frequently Bought Together