Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Scissors, universal

Bent, blunt/blunt, 180 mm
  RSGA109.185
 :   RSG
Scissors, universal
  RSGA109.185
 :  RSG
Scissors, universal
Scissors, universal

Bent, blunt/blunt, 180 mm

 
8,00 €
   
1 * 1 pezzi
 

Universal scissors with black plastic grip.

  • Autoclavable (max. 143 °C)
 
  • Type:
    Bent, blunt/blunt
  • Length:
    180 mm