3,5-Dinitroaniline
Catalog # EHERC12782200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Melting Pt:163 °C
Density:1.601 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1596
ADR:6.1,II
:183.12 g/mol
EINECS:210-567-8
CAS Number:618-87-1
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007263