Order Entry
Italy
ContactUsLinkComponent
2,6-Dinitroaniline
Toxic
Health
Environment
2,6-Dinitroaniline
Catalog # EHERC12782100
Supplier:  EHRENSTORFER
CAS Number:  
Toxic
Health
Environment
2,6-Dinitroaniline
Catalog # EHERC12782100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :183.12 g/mol
Merck Index:13,03299
Melting Pt:136 °C
Density:1.629 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-108-1
UN:1596
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:606-22-4
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007148