WHEATON® Diagnostic Vials
Supplier: DWK Life Sciences
Jump to:
About this item
These vials are manufactured from borosilicate glass that conforms to USP Type I requirements. The screw thread design eliminates the need for crimping tools.
- Alternative to serum bottles and vials
- Ideal for lyophilisation
- Fits with I-Loc™ closure or screw cap with thin flange stopper
- Heavy walled glass provides impact resistance
6 Options Available Below
Product Details & Documents
These vials are manufactured from borosilicate glass that conforms to USP Type I requirements. The screw thread design eliminates the need for crimping tools.
- Alternative to serum bottles and vials
- Ideal for lyophilisation
- Fits with I-Loc™ closure or screw cap with thin flange stopper
- Heavy walled glass provides impact resistance
Product Family Options
Clear vials
Product Information
- CapacityØ×HMaterialColourCap includedCap SizeAvailabilityPrice
- WHEAW219365Tubular vialsTubular vials5,00 ml22×38 mmBorosilicate glassClearNo20 - 400Log in for availability1 * 480 pezzi1 610,00 €
- WHEAW219366Tubular vialsTubular vials10,00 ml22×55 mmBorosilicate glassClearNo20 - 400Log in for availability1 * 480 pezzi1 710,00 €
Amber vials
Product Information
- CapacityØ×HMaterialColourCap includedCap SizeAvailabilityPrice
- WHEAW216375Tubular vialsTubular vials5,00 ml22×38 mmBorosilicate glassAmberNo20 - 400Log in for availability1 * 480 pezzi1 710,00 €
- WHEAW216376Tubular vialsTubular vials10,00 ml22×55 mmBorosilicate glassAmberNo20-400Log in for availability1 * 480 pezzi1 800,00 €
- WHEA219371Tubular vialsTubular vials10,00 ml25×61 mmBorosilicate glassAmberNo20 - 400Log in for availability1 * 480 pezzi1 950,00 €
- WHEAW216377Tubular vialsTubular vials20,00 ml30×52 mmBorosilicate glassAmberNo20 - 400Log in for availability1 * 480 pezzi1 860,00 €