Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Sanger's reagent 98%

Toxic
Health
Sanger's reagent 98%
Certificates
Sanger's reagent 98%

4 Options Available Below

Product Details & Documents
Synonyms:  1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene
  • CAS Number:70-34-8
  • Formula:C₆H₃FN₂O₄
  • MDL Number:MFCD00007056
  • Molecular Weight:186.1 g/mol
  • Merck Index:13,04199
  • Boiling Pt:296 °C
  • Melting Pt:23…26 °C
  • Density:1.48 g/cm³
  • Flash Pt:>112 °C
  • Storage Temperature:Ambient
  • EINECS:200-734-3
  • UN:2811
  • ADR:6.1,II

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to brown Liquid or low melting solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5675 to 1.5705 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Residue after ignition:
    ≤0.05 %
  • Suitability:
    (for precip. of Fe, etc.) Passes test
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 41,40 €
    41,40 €
    1 * 5 g
       41,40 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 75,60 €
    75,60 €
    1 * 25 g
       75,60 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 199,00 €
    199,00 €
    1 * 100 g
       199,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 770,00 €
    770,00 €
    1 * 500 g
       770,00 €