
3-Fluorobenzoic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:455-38-9
:
Formula:C₇H₅FO₂
MDL Number:MFCD00002489
:140.11 g/mol
Boiling Pt:251 °C (1013 hPa)
Melting Pt:121…125 °C
Storage Temperature:Ambient
EINECS:207-248-0
:GHS07
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:121 to 125 °C
- GC:≥98.5 %
- Titration with TBAH:≥98.5 %
- Solubility:(in boiling water) Soluble
- Related substances:Passes test
- Titration with NaOH:99.5 to 100.5 % (On dried substance)
- Chloride (Cl):≤200 ppm
Specification Test Results
Frequently Bought Together