
2-Fluorobenzoic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:445-29-4
:
Formula:C₇H₅FO₂
MDL Number:MFCD00002405
:140.11 g/mol
Boiling Pt:245 °C (1013 hPa)
Melting Pt:123…125 °C
Density:1,46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-158-1
:GHS07
Specification Test Results
- Appearance:White to light yellow Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:122 to 125 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together