1,4-Difluorobenzene 99+%
- :540-36-3
- :C₆H₄F₂
- :MFCD00000344
- :114.09 g/mol
- :13,03172
- :88 °C (1013 hPa)
- :–13 °C
- :1.17 g/cm³
- :2 °C
- :Ambient
- :208-742-9
- :1993
- :3,II
- :540-36-3
- :C₆H₄F₂
- :MFCD00000344
- :114.09 g/mol
- :13,03172
- :88 °C (1013 hPa)
- :–13 °C
- :1.17 g/cm³
- :2 °C
- :Ambient
- :208-742-9
- :1993
- :3,II
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4420 (20°C, 589 nm)
- GC:≥99.0 %
Product Family Options
- Pack typePk



