Order Entry
Italy
ContactUsLinkComponent
2,4-Dichlorophenol 99%
Toxic
Corrosive
Environment
2,4-Dichlorophenol 99%
Certificates
undefined
Toxic
Corrosive
Environment
2,4-Dichlorophenol 99%

Specification Test Results

  • Appearance:
    White to beige Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    41 to 44 °C
  • Water:
    ≤0.3 %
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    +9.0° to +11.0° (20 °C, 589 nm) (C=2 in methanol)

Specification Test Results

Related Information

Frequently Bought Together