Order Entry
Italy
ContactUsLinkComponent
3,4-Dichlorophenol for synthesis, Sigma-Aldrich®
Irritant
3,4-Dichlorophenol for synthesis, Sigma-Aldrich®
Catalog # 8.20450.0100
Supplier:  Merck
CAS Number:  
Irritant
3,4-Dichlorophenol for synthesis, Sigma-Aldrich®
Catalog # 8.20450.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    9260 mg/l (25 °C) (experimental)
  • Melting Point:
    66 - 68 °C
  • Molar Mass:
    163 g/mol
  • Boiling Point:
    146 °C (1013 hPa)

Specification Test Results

About this item

 :
Boiling Pt:247 °C (1013 hPa)
Melting Pt:66…68 °C
Density:1.6 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2020
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
 :163 g/mol
EINECS:202-450-5
 :GHS07
CAS Number:95-77-2
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002258