

1,5-Dichloro-2,4-dinitrobenzene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3698-83-7
:
Formula:C₆H₂Cl₂N₂O₄
MDL Number:MFCD00024186
:237 g/mol
Boiling Pt:135…155 °C (0.5 mmHg)
Melting Pt:98…103 °C
Storage Temperature:Ambient
EINECS:223-027-1
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together