Order Entry
Italy
ContactUsLinkComponent
1,3-Dichloro-4-nitrobenzene for synthesis
Toxic
Environment
1,3-Dichloro-4-nitrobenzene for synthesis
Catalog # 8.20446.1000
Supplier:  Merck
CAS Number:  
Toxic
Environment
1,3-Dichloro-4-nitrobenzene for synthesis
Catalog # 8.20446.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
    188 g/l (20 °C)
  • Melting Point:
    28 - 31 °C
  • Molar Mass:
    192 g/mol
  • Boiling Point:
    259 °C
  • Vapor Pressure:
    0.013 hPa (20 °C)
  • Flash Point:
    152 °C
  • Density:
    1.51 g/cm3
  • Ignition Point:
    500 °C

Specification Test Results

About this item

 :
 :192 g/mol
Boiling Pt:258 °C (1013 hPa)
Melting Pt:28…31 °C
Density:1.51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-248-3
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS09
CAS Number:611-06-3
Formula:C₆H₃Cl₂NO₂
MDL Number:MFCD00007071