Order Entry
Italy
ContactUsLinkComponent
3-Chloro-4-fluoroaniline 98%
Irritant
3-Chloro-4-fluoroaniline 98%
Certificates
undefined
Irritant
3-Chloro-4-fluoroaniline 98%

Specification Test Results

  • Appearance:
    Beige to brown Crystalline solid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    42 to 47 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together