Order Entry
Italy
ContactUsLinkComponent
1,3-Dichloro-4-fluorobenzene ≥99%
  A17953.18
 :  
1,3-Dichloro-4-fluorobenzene ≥99%
  A17953.18
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368
 :1435-48-9
 :
 :C₆H₃Cl₂F
 :MFCD00000580
 :164.99 g/mol
 :172…174 °C
 :–23 °C
 :1.409
 :79 °C (174 °F)
 :Ambient
 :406-160-1
 :2810
 :6.1,III
 :GHS08
 :GHS07
 :GHS09