4-Chlorobenzoic acid
EHERC11391000
:
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
:
:156.57 g/mol
:13,02144
:287 °C (1013 hPa)
:123 °C
:1.535 g/cm³ (20 °C)
:238 °C
:Ambient
:200-805-9
:GHS07
:74-11-3
:C₇H₅ClO₂
:MFCD00002531