Order Entry
Italy
ContactUsLinkComponent
 
 
2-Chlorobenzoic acid for synthesis, Sigma-Aldrich®
  8.00463.0500
 :  Merck
 :  
 
2-Chlorobenzoic acid for synthesis, Sigma-Aldrich®
  8.00463.0500
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Solubility:
    2.1 g/l (25 °C)
  • Melting Point:
    139 - 142 °C
  • Molar Mass:
    156.57 g/mol
  • Bulk Density:
    600 kg/m3
  • Boiling Point:
    284 - 286 °C (1013 hPa)
  • Vapor Pressure:
    0.001 hPa (25 °C)
  • Density:
    1.544 g/cm3 (20 °C)
  • pH:
    3.4 (0.2 g/l, H2O, 20 °C)
  • Ignition Point:
    530 °C

 

 

 :
 :156.57 g/mol
 :13,02143
 :285 °C (1013 hPa)
 :139…142 °C
 :1.544 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :173 °C
 :Ambient
 :204-285-4
 :GHS07
 :118-91-2
 :C₇H₅ClO₂
 :MFCD00002412