
2,4-Dichlorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50-84-0
:
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002414
:191.01 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:159…164 °C
Storage Temperature:Ambient
EINECS:200-067-8
:GHS07
Related Information
Certificates
Search by Lot Numbermsds_ALFAA12372
Scheda di sicurezza
Organic Building Blocks and Intermediates
Note applicative
Frequently Bought Together