

2,4-Dichlorobenzaldehyde per sintesi, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:874-42-0
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00003305
:175.01 g/mol
Boiling Pt:233 °C (1013 hPa)
Melting Pt:68…71 °C
Density:1,33 g/cm³ (20 °C)
Storage Temperature:Ambiente
EINECS:212-861-1
ADR:9,III
:M-Clarity™ quality level = MQ200
UN:3077
:GHS07
:GHS09
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:68 - 71 °C
- Molar Mass:175.01 g/mol
- Bulk Density:700 kg/m3
- Boiling Point:233 °C (1013 hPa)
- Flash Point:135 °C
- Ignition Point:440 °C
Specification Test Results
Frequently Bought Together