Order Entry
Italy
ContactUsLinkComponent
Chlorobenzene 99+%, pure
Irritant
Environment
Flammable
Chlorobenzene 99+%, pure
Certificates
undefined
Irritant
Environment
Flammable
Chlorobenzene 99+%, pure

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5230 to 1.5250 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA

Specification Test Results

Related Information

Frequently Bought Together