Order Entry
Italy
ContactUsLinkComponent
Salicylic acid 99+%
Salicylic acid 99+%
Certificates
undefined
Salicylic acid 99+%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    158 to 161 °C
  • Titration with NaOH:
    99.0 to 101.0 %

Specification Test Results

Related Information

Frequently Bought Together