
3,5-Dinitrosalicylic acid for synthesis, Sigma-Aldrich®
8.00141.2500
: Merck
:
- Pack type:Plastic bottle for solids
- Pk:2,5 kg
- Seq:4076
- Solubility:(20 °C) soluble
- Melting Point:170 - 174 °C
- Molar Mass:228.12 g/mol
- Bulk Density:400 kg/m3
- pH:1.3 - 1.8 (10 g/l, H2O, 20 °C)
- Synonyms:3,5-Dinitro-2-hydroxybenzoic acid, 3,5-Dinitrosalicylic acid
- Hill Formula:C7H4N2O7
- Chemical Formula:2-(OH)-3,5-(NO2)2C6H2COOH
:
:228.12 g/mol
:170…174 °C
:Ambient
:8,III
:M-Clarity™ quality level = MQ200
:210-204-3
:GHS07
:609-99-4
:C₇H₄N₂O₇
:MFCD00007104