
3,5-Diaminobenzoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:535-87-5
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007807
:152.15 g/mol
Merck Index:13,02997
Boiling Pt:546 °C (1013 hPa)
Melting Pt:235 °C (dec.)
Density:1,45 g/cm³ (20 °C)
EINECS:208-621-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to grey Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Titration with TBAH:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together