Order Entry
Italy
Orders LinkContactUsLinkComponent
Micro-Ostwald viscometers
Micro-Ostwald viscometers
Supplier:   SI Analytics
Micro-Ostwald viscometers
Supplier:  SI Analytics
Certificates
Micro-Ostwald viscometers
15 Options Available Below
Product Details & Documents
These Micro-Ostwald viscometers are available as calibrated for either manual or automatic measurements and uncalibrated models.
Suitable for measurements of small liquid quantities with extreme formation of foam.
Product Family Options
Calibrated for manual measurements
Product Information
  • Range
    Constant
    Length
    Ø int.
    Availability
    Price
  • 0,4 - 6 mm²/s
    0,01 K
    290 mm
    0,43 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
  • 1,2 - 18 mm²/s
    0,03 K
    290 mm
    0,60 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
  • 4 - 60 mm²/s
    0,1 K
    290 mm
    0,77 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
  • 12 - 180 mm²/s
    0,3 K
    290 mm
    1,00 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
  • 40 - 800 mm²/s
    1 K
    290 mm
    1,36 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
Calibrated for automatic measurements
Product Information
  • Range
    Constant
    Length
    Ø int.
    Availability
    Price
  • 0,4 - 6 mm²/s
    0,01 K
    290 mm
    0,43 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
  • 1,2 - 18 mm²/s
    0,03 K
    290 mm
    0,60 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
  • 4 - 60 mm²/s
    0,1 K
    290 mm
    0,77 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
  • 12 - 180 mm²/s
    0,3 K
    290 mm
    1,00 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
  • 40 - 800 mm²/s
    1 K
    290 mm
    1,36 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 455,00 €
    455,00 €
    1 * 1 pezzi
       455,00 €
Uncalibrated
Product Information
  • Range
    Constant
    Length
    Ø int.
    Availability
    Price
  • 0,4 - 6 mm²/s
    290 mm
    0,43 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 285,00 €
    285,00 €
    1 * 1 pezzi
       285,00 €
  • 1,2 - 18 mm²/s
    290 mm
    0,60 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 285,00 €
    285,00 €
    1 * 1 pezzi
       285,00 €
  • 4 - 60 mm²/s
    290 mm
    0,77 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 285,00 €
    285,00 €
    1 * 1 pezzi
       285,00 €
  • 12 - 180 mm²/s
    290 mm
    1,00 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 285,00 €
    285,00 €
    1 * 1 pezzi
       285,00 €
  • 40 - 800 mm²/s
    290 mm
    1,36 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 285,00 €
    285,00 €
    1 * 1 pezzi
       285,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Polyamide bracket holder for routine viscometers, Cannon-Fenske reverse flow viscometers and Ostwald viscometers
    Polyamide bracket holder for routine viscometers, Cannon-Fenske reverse flow viscometers and Ostwald viscometers
    442-1233
    Log in for availability
    1 * 1 pezzi - 83,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone ring (i.Ø - o.Ø = 6 - 16 mm)
    Silicone ring (i.Ø - o.Ø = 6 - 16 mm)
    442-1235
    Log in for availability
    1 * 1 pezzi - 16,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone ring (i.Ø - o.Ø = 4 - 10 mm)
    Silicone ring (i.Ø - o.Ø = 4 - 10 mm)
    442-1234
    Log in for availability
    1 * 1 pezzi - 13,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone ring (i.Ø - o.Ø = 8 - 16 mm)
    Silicone ring (i.Ø - o.Ø = 8 - 16 mm)
    442-1236
    Log in for availability
    1 * 1 pezzi - 13,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone ring (i.Ø - o.Ø = 10 - 16 mm)
    Silicone ring (i.Ø - o.Ø = 10 - 16 mm)
    442-1237
    Log in for availability
    1 * 1 pezzi - 14,80 €