Order Entry
Italy
Orders LinkContactUsLinkComponent
Filter funnels, DURAN®
Filter funnels, DURAN®
Supplier:   DWK Life Sciences
Filter funnels, DURAN®
Supplier:  DWK Life Sciences
Certificates
Filter funnels, DURAN®
Filter funnels, DURAN®
2 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filter plate.
Product Family Options
Product Information
  • Capacity
    Length
    Plate Ø
    Stem length
    Porosity
    Stem O-Ø
    Availability
    Price
  • 25 ml
    100 mm
    25 mm
    55 mm
    3
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 52,40 €
    52,40 €
    1 * 1 pezzi
       52,40 €
  • 25 ml
    100 mm
    25 mm
    55 mm
    4
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 58,60 €
    58,60 €
    1 * 1 pezzi
       58,60 €
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..