Benzoic acid methyl ester, Supelco®
Catalog # 18344-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:136.15 g/mol
Merck Index:13,06052
Boiling Pt:199.6 °C (1013 hPa)
Melting Pt:–12 °C
Density:1.0888 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:83 °C
Storage Temperature:Ambient
EINECS:202-259-7
:GHS07
CAS Number:93-58-3
Formula:C₆H₅COOCH₃
MDL Number:MFCD00008421