
2,3,4-Trihydroxybenzaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2144-08-3
:
Formula:C₇H₆O₄
MDL Number:MFCD00003325
:154.12 g/mol
Boiling Pt:332 °C (1013 hPa)
Melting Pt:159…162 °C
Density:1,054 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:218-404-2
:GHS07
Specification Test Results
- Appearance:Beige to light brown Powder
- Infrared spectrum:Conforms
- Melting point:159 to 162 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together