Order Entry
Italy
ContactUsLinkComponent
2,3-Dimethoxybenzaldehyde 97%
Irritant
2,3-Dimethoxybenzaldehyde 97%
Certificates
undefined
Irritant
2,3-Dimethoxybenzaldehyde 97%

Specification Test Results

  • Appearance:
    Off-white to beige Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    48 to 53 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together