Standing cylinders, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- Good temperature resistance
- Sturdy
Universal standing cylinder without graduation, e.g. for mixing several components.
Frequently Bought Together