4-Bromophenol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-41-2
:
Formula:C₆H₅BrO
MDL Number:MFCD00002313
:173.01 g/mol
Merck Index:14,1428
Boiling Pt:235…238 °C
Melting Pt:62…66 °C
Density:1.840
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:203-394-4
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together