Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dibromophenol
2,4-Dibromophenol
2,4-Dibromophenol
EHERC12241000
:
EHRENSTORFER
:
2,4-Dibromophenol
EHERC12241000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
251.91 g/mol
:
238…239 °C (1013 hPa)
:
38 °C
:
2.07 g/cm³ (20 °C)
:
210-436-5
:
2811
:
6.1,II
:
615-58-7
:
C₆H₄Br₂O
:
MFCD00002149