(±)-1-Butoxy-2-propanol ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5131-66-8
:
Formula:C₇H₁₆O₂
MDL Number:MFCD00021904
:132.2 g/mol
Boiling Pt:170°C
Flash Pt:62°C
EINECS:225-878-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together