Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Dimethylnitrosoamine 200 µg/ml in methanol, Supelco®
Organic reference standard, Dimethylnitrosoamine 200 µg/ml in methanol, Supelco®
Catalog # 48670.
Supplier:  Merck
CAS Number:  
Organic reference standard, Dimethylnitrosoamine 200 µg/ml in methanol, Supelco®
Catalog # 48670.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    62-75-9
  • Description:
    Dimethylnitrosoamine 200 µg/ml in methanol
  • Formula:
    C₂H₆N₂O
  • Description:
    Dimethylnitrosoamine

Specifications

About this item