Organic reference standard, 3,5-Xylenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12735000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:108-68-9
- Description:3,5-Xylenol 100 µg/ml in methanol
- Formula:C₈H₁₀O
- Description:3,5-Xylenol