Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, 2,4',5-Trichlorobiphenyl (PCB No. 31) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, 2,4',5-Trichlorobiphenyl (PCB No. 31) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20003100CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,4',5-Trichlorobiphenyl (PCB No. 31) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20003100CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    16606-02-3
  • Description:
    2,4',5-Trichlorobiphenyl (PCB No. 31) 10 µg/ml in cyclohexane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,4',5-Trichlorobiphenyl (PCB No. 31)

Specifications

About this item