Organic reference standard, 2,4,6-Trichlorobiphenyl (PCB No. 30) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL20003000CY
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:35693-92-6
- Description:2,4,6-Trichlorobiphenyl (PCB No. 30) 10 µg/ml in cyclohexane
- Formula:C₁₂H₇Cl₃
- Description:2,4,6-Trichlorobiphenyl (PCB No. 30)