Organic reference standard, m-Xylene 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL17945100ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:108-38-3
- Description:m-Xylene 10 µg/ml in methanol
- Formula:C₈H₁₀
- Description:m-Xylene