Pesticide reference standard, 1,2,3-Trichlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17722300CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:87-61-6
- Description:1,2,3-Trichlorobenzene 10 µg/ml in cyclohexane
- Formula:C₆H₃Cl₃
- Description:1,2,3-Trichlorobenzene