Order Entry
Italy
ContactUsLinkComponent
 
Ethyl pelargonate
Catalog # EHERC15623130
Supplier:  EHRENSTORFER
CAS Number:  
Ethyl pelargonate
Catalog # EHERC15623130
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :186.29 g/mol
EINECS:204-615-7
CAS Number:123-29-5
Formula:C₁₁H₂₂O₂
MDL Number:MFCD00009570