Order Entry
Italy
ContactUsLinkComponent
 
Irritant
5-Bromo-2-fluoro-3-pyridineboronic acid ≥98%
Catalog # L19915.06
CAS Number:  
Irritant
5-Bromo-2-fluoro-3-pyridineboronic acid ≥98%
Catalog # L19915.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :219.8 g/mol
Melting Pt:>300 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:501435-91-2
Formula:C₅H₄BBrFNO₂
MDL Number:MFCD04039317